Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD2D Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP103357B
Description
JMJD2D Polyclonal specifically detects JMJD2D in Human samples. It is validated for Western Blot, ICC/IF (Negative).Specifications
JMJD2D | |
Polyclonal | |
Western Blot, ICC/IF (Negative) | |
EC 1.14.11, EC 1.14.11.-, FLJ10251, JHDM3D, JmjC domain-containing histone demethylation protein 3D, JMJD2Dlysine-specific demethylase 4D, jumonji domain containing 2D, Jumonji domain-containing protein 2D, lysine (K)-specific demethylase 4D, MGC141909 | |
Rabbit | |
55 kDa | |
0.1 mL | |
Transcription Factors and Regulators | |
55693 | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry | |
Biotin | |
PBS with 0.05% Sodium Azide | |
KDM4D | |
Synthetic peptide made to a C-terminal portion of the human JMJD2D protein (within residues 460-520). [Swiss-Prot# Q6B0I6] | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction