Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD2D Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA511148

Catalog No. PIPA511148


Only null left
Add to Cart

Description

Description

JMJD2D Polyclonal Antibody for Western Blot

JMJD2D belongs to JMJD2 family that is consist of four cancer-associated genes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

JMJD2D
Polyclonal
Unconjugated
KDM4D
FLJ10251; JHDM3D; JmjC domain-containing histone demethylation protein 3D; JMJD2D; jumonji domain containing 2D; jumonji domain-containing protein 2D; KDM4D; lysine (K)-specific demethylase 4D; lysine demethylase 4D; lysine-specific demethylase 4D
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
55693
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1.8 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6B0I6
KDM4D
KLH conjugated synthetic peptide between 484-516 amino acids from the C-terminal region of human JMJD2D.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.