Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2D Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992654
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2654 10 μL
50-199-2672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2654 Supplier Active Motif Supplier No. 39248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JMJD2D belongs to JMJD2 family that is consist of four cancer-associated genes.

Specifications

Antigen JMJD2D
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene KDM4D
Gene Accession No. Q6B0I6
Gene Alias FLJ10251; Jhdm3d; JmjC domain-containing histone demethylation protein 3D; Jmjd2d; jumonji domain containing 2D; Jumonji domain-containing protein 2D; Kdm4d; lysine (K)-specific demethylase 4D; lysine demethylase 4D; Lysine-specific demethylase 4D
Gene Symbols KDM4D
Host Species Rabbit
Immunogen This JMJD2D antibody was raised against a peptide corresponding to amino acids 337-359 of human JMJD2D.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55693
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.