Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD3 / KDM6B Monoclonal Antibody (67-A2), Active Motif™
SDP

Catalog No. 501993363
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3363 100 μg
50-199-3364 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3363 Supplier Active Motif Supplier No. 61387
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Covalent modification of histones plays critical role in regulating chromatin structure and transcription. While most covalent histone modifications are reversible, only recently has it been established that methyl groups are subject to enzymatic removal from histones. A family of novel JmjC domain-containing histone demethylation (JHDM) enzymes have been identified that perform this specific function. Histone demethylation by JHDM proteins requires cofactors Fe(II) and alpha-ketoglutarate. Family members include JHDM1 (demethylating histone 3 at lysine 36), and JHDM2A as well as JMJD2CH3K9 (both of which demethylate histone 3 at lysine 9). Contributions of histone demethylase activity to tumor development, decreases in cell proliferation, and hormone-dependent transcriptional activation have been observed.

Specifications

Antigen JMJD3 / KDM6B
Applications Western Blot
Classification Monoclonal
Clone 67-A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene KDM6B
Gene Accession No. O15054, Q5NCY0
Gene Alias 1700064E03Rik; BC038313; JmjC domain-containing protein 3; Jmjd3; jumonji domain containing 3; jumonji domain containing 3, histone lysine demethylase; jumonji domain-containing 3; Jumonji domain-containing protein 3; KDM1 lysine (K)-specific demethylase 6B; Kdm6b; Kiaa0346; lysine (K)-specific demethylase 6B; Lysine demethylase 6B; Lysine-specific demethylase 6B; mKIAA0346
Gene Symbols KDM6B
Host Species Mouse
Immunogen This antibody was raised against a recombinant protein corresponding to amino acids 1028-1684 of human JMJD3 / kDM6B.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216850, 23135
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.