Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113675 Supplier Invitrogen™ Supplier No. PA5113675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRAVDPPGAR AAREAFALGG LSCAGAWSSC PPHPPPRSAW LPGGRCSASI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

Covalent modification of histones plays critical role in regulating chromatin structure and transcription. While most covalent histone modifications are reversible, only recently has it been established that methyl groups are subject to enzymatic removal from histones. A family of novel JmjC domain-containing histone demethylation (JHDM) enzymes have been identified that perform this specific function. Histone demethylation by JHDM proteins requires cofactors Fe(II) and alpha-ketoglutarate. Family members include JHDM1 (demethylating histone 3 at lysine 36), and JHDM2A as well as JMJD2CH3K9 (both of which demethylate histone 3 at lysine 9). Contributions of histone demethylase activity to tumor development, decreases in cell proliferation, and hormone-dependent transcriptional activation have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KDM6B
Gene Accession No. O15054
Gene Alias 1700064E03Rik; BC038313; jmjC domain-containing protein 3; JMJD3; jumonji domain containing 3; jumonji domain containing 3, histone lysine demethylase; jumonji domain-containing 3; jumonji domain-containing protein 3; KDM1 lysine (K)-specific demethylase 6B; Kdm6b; KIAA0346; lysine (K)-specific demethylase 6B; lysine demethylase 6B; Lysine-specific demethylase 6B; mKIAA0346
Gene Symbols KDM6B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human JMJD3 (aa 2-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23135
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.