Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    JMJD4 Antibody (JMJD4/32b/H12), mFluor Violet 450 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250425MFV450
Description
JMJD4 Monoclonal antibody specifically detects JMJD4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, KnockDownSpecifications
| JMJD4 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, KnockDown | |
| JMJD4/32b/H12 | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Knockdown Validated | |
| FLJ12517, jumonji domain containing 4, jumonji domain-containing protein 4 | |
| Recombinant protein | |
| 0.1 mL | |
| Epigenetics | |
| 65094 | |
| Store at 4C in the dark. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            