Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JMJD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520805
Description
A suggested positive control is human spleen tissue lysate. PA5-20805 can be used with blocking peptide PEP-0919.
Covalent modification of histones plays critical role in regulating chromatin structure and transcription. While most covalent histone modifications are reversible, only recently has it been established that methyl groups are subject to enzymatic removal from histones. A family of novel JmjC domain-containing histone demethylation (JHDM) enzymes have been identified that perform this specific function. Histone demethylation by JHDM proteins requires cofactors Fe(II) and alpha-ketoglutarate. Family members include JHDM1 (demethylating histone 3 at lysine 36), and JHDM2A as well as JMJD2CH3K9 (both of which demethylate histone 3 at lysine 9). Contributions of histone demethylase activity to tumor development, decreases in cell proliferation, and hormone-dependent transcriptional activation have been observed.
Specifications
JMJD4 | |
Polyclonal | |
Unconjugated | |
Jmjd4 | |
2-oxoglutarate- and Fe(II)-dependent oxygenase; 2-oxoglutarate and iron-dependent oxygenase JMJD4; 6430559I23; C4 lysyl hydroxylase; jmjC domain-containing protein 4; JMJD4; jumonji domain containing 4; jumonji domain-containing protein 4; Lysyl-hydroxylase JMJD4; RGD1307186 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
287359, 65094 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9H9V9 | |
Jmjd4 | |
A 17 amino acid peptide from near the amino terminus of human JMJD4. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction