Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596889 Supplier Invitrogen™ Supplier No. PA596889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRAGPEPQAL AGQKRGALRL LVPRLVLTVS APAEVRRRVL RPVLSWMDRE TRALADSHFR GLGVDVPGVG QAPGRVAFVS EPGAFSYADF VRGFLLPNLP CVFSSAFTQG WGSRRRWVTP AGRPDFDHLL RTYGDVVVPV ANCGVQEYNS NPKEHMTLRD YITYWKEYIQ AGYSSPRGCL YLKDWHLCRD FPVEDVFTLP VYFSSDWLNE FWDALDVDDY RFVYAGPAGS WSPFHADIFR SFSWSVNVCG RKKWLLFPPG QEEALRDRHG NLPYDVTSPA LCDTHLHPRN QLAGPPLEIT QEAGEMVFVP SGWHHQVHNL DDTISINHNW VNGFNLANMW RFLQQELCAV QEEVSEWRDS MPDWHHHCQV IMRSCSGINF EEFYHFLKVI AEKRLLVLRE AAAEDGAGLG FEQAAFDVGR ITEVLASLVA HPDFQRVDTS AFSPQPKELL QQLREAVDAA AAP; Positive Samples: 293T, HepG2, NCI-H460, BT-474, MCF7, Mouse liver, Mouse brain, Rat liver.

Covalent modification of histones plays critical role in regulating chromatin structure and transcription. While most covalent histone modifications are reversible, only recently has it been established that methyl groups are subject to enzymatic removal from histones. A family of novel JmjC domain-containing histone demethylation (JHDM) enzymes have been identified that perform this specific function. Histone demethylation by JHDM proteins requires cofactors Fe(II) and alpha-ketoglutarate. Family members include JHDM1 (demethylating histone 3 at lysine 36), and JHDM2A as well as JMJD2CH3K9 (both of which demethylate histone 3 at lysine 9). Contributions of histone demethylase activity to tumor development, decreases in cell proliferation, and hormone-dependent transcriptional activation have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Jmjd4
Gene Accession No. Q8BFT6, Q9H9V9
Gene Alias 2-oxoglutarate- and Fe(II)-dependent oxygenase; 2-oxoglutarate and iron-dependent oxygenase JMJD4; 6430559I23; C4 lysyl hydroxylase; jmjC domain-containing protein 4; JMJD4; jumonji domain containing 4; jumonji domain-containing protein 4; Lysyl-hydroxylase JMJD4; RGD1307186
Gene Symbols Jmjd4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-463 of human JMJD4 (NP_0753832).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 194952, 287359, 65094
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.