Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30472720UL
Description
JMJD4 Polyclonal specifically detects JMJD4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
JMJD4 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
65094 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
FLJ12517, jumonji domain containing 4, jumonji domain-containing protein 4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-463 of human JMJD4 (NP_075383.2). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction