Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD5 (KDM8) Monoclonal Antibody (OTI5E8), TrueMAB™, OriGene
SDP

Catalog No. 502543990
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3990 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3990 Supplier OriGene Supplier No. CF810952
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JMJD5 (KDM8) Monoclonal antibody specifically detects JMJD5 (KDM8) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen JMJD5 (KDM8)
Applications Western Blot
Classification Monoclonal
Clone OTI5E8
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene SPOCK2
Gene Accession No. Q92563, Q9ER58
Gene Alias 3110005O21Rik; arginyl C3-hydroxylase KDM8; Bifunctional peptidase and arginyl-hydroxylase JMJD5; bifunctional peptidase and arginyl-hydroxylase JMJD5; jmjC domain-containing protein 5; hypothetical protein FLJ13798; jmjC domain-containing protein 5; Jmjd5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; Kdm8; L-arginine (3R)-hydroxylase KDM8; LOW QUALITY PROTEIN: lysine-specific demethylase 8; lysine (K)-specific demethylase 8; lysine demethylase 8; lysine-specific demethylase 8; RGD1304823; zgc:173863; zgc:91962
Host Species Mouse
Immunogen Full length human recombinant protein of human SPOCK2 (NP_055582) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361840, 94214, 9806
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.