Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100205 Supplier Invitrogen™ Supplier No. PA5100205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KDM8.

This gene likely encodes a histone lysine demethylase. Studies of a similar protein in mouse indicate a potential role for this protein as a tumor suppressor. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kdm8
Gene Accession No. Q497B8, Q8N371, Q9CXT6
Gene Alias 3110005O21Rik; arginyl C3-hydroxylase KDM8; Bifunctional peptidase and arginyl-hydroxylase JMJD5; bifunctional peptidase and arginyl-hydroxylase JMJD5; jmjC domain-containing protein 5; hypothetical protein FLJ13798; jmjC domain-containing protein 5; Jmjd5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; Kdm8; L-arginine (3R)-hydroxylase KDM8; LOW QUALITY PROTEIN: lysine-specific demethylase 8; lysine (K)-specific demethylase 8; lysine demethylase 8; lysine-specific demethylase 8; RGD1304823; zgc:173863; zgc:91962
Gene Symbols Kdm8
Host Species Rabbit
Immunogen A synthesized peptide derived from human KDM8(Accession Q8N371), corresponding to amino acid residues I291-Y341.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308976, 77035, 79831
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.