Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988231
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8231 Supplier Bioss Supplier No. BS8047R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene likely encodes a histone lysine demethylase. Studies of a similar protein in mouse indicate a potential role for this protein as a tumor suppressor. Alternatively spliced transcript variants have been described.

Specifications

Antigen JMJD5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Kdm8
Gene Accession No. Q8N371
Gene Alias 3110005O21Rik; Bifunctional peptidase and arginyl-hydroxylase JMJD5; BOS_22772; JmjC domain-containing protein 5; JMJD5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; KDM8; L-arginine (3R)-hydroxylase KDM8; lysine (K)-specific demethylase 8; lysine demethylase 8; Lysine-specific demethylase 8; RGD1304823; zgc:173863; zgc:91962
Gene Symbols Kdm8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human JMJD5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79831
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.