Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMJD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111481
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111481 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111481 Supplier Invitrogen™ Supplier No. PA5111481
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STPRELIKVT RDEGGNQQDE AITWFNVIYP RTQLPTWPPE FKPLEILQKP GETVFVPGGW WHVVLNLDTT IAITQNFASS TNFPVVWHKT VRGRPKLSRK WYRILKQEHP ELAVLA Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene encodes a nuclear protein with a JmjC domain. JmjC domain-containing proteins are predicted to function as protein hydroxylases or histone demethylases. This protein was first identified as a putative phosphatidylserine receptor involved in phagocytosis of apoptotic cells; however, subsequent studies have indicated that it does not directly function in the clearance of apoptotic cells, and questioned whether it is a true phosphatidylserine receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JMJD6
Gene Accession No. Q6NYC1
Gene Alias 5730436I23Rik; apoptotic cell clearance receptor PtdSerR; arginine demethylase and lysine hydroxylase; bifunctional arginine demethylase and lysyl-hydroxylase JMJD6; D11Ertd195e; Histone arginine demethylase JMJD6; jmjC domain-containing protein 6; Jmjd6; jumonji domain containing 6; jumonji domain-containing protein 6; KIAA0585; Lysyl-hydroxylase JMJD6; mKIAA0585; Peptide-lysine 5-dioxygenase JMJD6; phosphatidylserine receptor; Protein PTDSR; PSR; PtdSerR; Ptdsr; PTDSR1
Gene Symbols JMJD6
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human JMJD6. Recombinant protein control fragment (Product #RP-105123).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23210
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.