Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD6 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993448
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3448 10 μL
50-199-3447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3448 Supplier Active Motif Supplier No. 61494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a nuclear protein with a JmjC domain. JmjC domain-containing proteins are predicted to function as protein hydroxylases or histone demethylases. This protein was first identified as a putative phosphatidylserine receptor involved in phagocytosis of apoptotic cells; however, subsequent studies have indicated that it does not directly function in the clearance of apoptotic cells, and questioned whether it is a true phosphatidylserine receptor. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen JMJD6
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene JMJD6
Gene Accession No. Q6NYC1
Gene Alias 5730436I23Rik; apoptotic cell clearance receptor PtdSerR; arginine demethylase and lysine hydroxylase; Bifunctional arginine demethylase and lysyl-hydroxylase JMJD6; D11Ertd195e; Histone arginine demethylase JMJD6; JmjC domain-containing protein 6; Jmjd6; jumonji domain containing 6; Jumonji domain-containing protein 6; Kiaa0585; Lysyl-hydroxylase JMJD6; mKIAA0585; Peptide-lysine 5-dioxygenase JMJD6; Phosphatidylserine receptor; Protein PTDSR; PSR; PtdSerR; Ptdsr; PTDSR1
Gene Symbols JMJD6
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the N-terminal region of human JMJD6.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23210
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.