Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD6/PSR Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7202681 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092550 20 μL
NB092549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092550 Supplier Novus Biologicals Supplier No. NBP30344420UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JMJD6/PSR Polyclonal specifically detects JMJD6/PSR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen JMJD6/PSR
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias bifunctional arginine demethylase and lysyl-hydroxylase JMJD6, EC 1.14.11, EC 1.14.11.-, Histone arginine demethylase JMJD6, JmjC domain-containing protein 6, jumonji domain containing 6, Jumonji domain-containing protein 6, KIAA0585PSR, Lysyl-hydroxylase JMJD6, Peptide-lysine 5-dioxygenase JMJD6, Phosphatidylserine receptorPTDSR, Protein PTDSR, PTDSR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human JMJD6/PSR (NP_055982.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Epigenetics, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 23210
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.