Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMY Antibody (HMY117), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250457G
Description
JMY Monoclonal specifically detects JMY in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
JMY | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
JMY | |
Synthetic peptide from the c terminus of the peptide sequence, identical for both isoform 1 and 2 | |
0.1 mL | |
Apoptosis | |
133746 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry | |
HMY117 | |
Western Blot, Immunohistochemistry | |
FLJ37870, junction mediating and regulatory protein, p53 cofactor, junction-mediating and regulatory protein, junction-mediating and -regulatory protein, MGC163496, WAS protein homology region 2 domain containing 1-like 3, WHDC1L3 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction