Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMY Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130464JF646
Description
JMY Polyclonal antibody specifically detects JMY in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
JMY | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ37870, junction mediating and regulatory protein, p53 cofactor, junction-mediating and regulatory protein, junction-mediating and -regulatory protein, MGC163496, WAS protein homology region 2 domain containing 1-like 3, WHDC1L3 | |
The immunogen for this product maps to a region between residue 1 and 50 of human junction mediating and regulatory protein using the numbering given in entry NP_689618.3 (GeneID 133746). | |
0.1 mL | |
Apoptosis | |
133746 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction