Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMY Antibody, Novus Biologicals™
SDP

Catalog No. NB260496 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260496 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB260496 Supplier Novus Biologicals Supplier No. NBP323402
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JMY Polyclonal antibody specifically detects JMY in Human, Mouse, Rat, Chicken, Xenopus samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen JMY
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200
Formulation PBS, 1 mg/ml BSA and 50% glycerol
Gene Alias FLJ37870, junction mediating and regulatory protein, p53 cofactor, junction-mediating and regulatory protein, junction-mediating and -regulatory protein, MGC163496, WAS protein homology region 2 domain containing 1-like 3, WHDC1L3
Host Species Rabbit
Immunogen JMY synthetic peptide (coupled to carrier protein) corresponds to amino acids in the C-terminal region of mouse JMY. This sequence is highly conserved in human, rat, frog, and zebrafish JMY proteins, and has low homology to other WASP family members. [UniProt# Q9QXM1]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 133746
Target Species Human, Mouse, Rat, Chicken, Xenopus species
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.