Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JMY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560317 Supplier Invitrogen™ Supplier No. PA560317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSSVRIVSAS GTVSEEIEVL EMVKEDEAPL ALSDAEQPPP ATELESPAEE CSWAGLFSFQ DLRAVHQQLC SVNSQLEPCL PVFPEEPSGM WTVLF Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.

JMY acts both as a nuclear p53/TP53-cofactor and a cytoplasmic regulator of actin dynamics depending on conditions. In nucleus, acts as a cofactor that increases p53/TP53 response via its interaction with p300/EP300. Increases p53/TP53-dependent transcription and apoptosis, suggesting an important role in p53/TP53 stress response such as DNA damage. In cytoplasm, acts as a nucleation-promoting factor for both branched and unbranched actin filaments. Activates the Arp2/3 complex to induce branched actin filament networks. Also catalyzes actin polymerization in the absence of Arp2/3, creating unbranched filaments. Contributes to cell motility by controlling actin dynamics. May promote the rapid formation of a branched actin network by first nucleating new mother filaments and then activating Arp2/3 to branch off these filaments. The p53/TP53-cofactor and actin activator activities are regulated via its subcellular location.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMY
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JMY
Gene Accession No. Q8N9B5
Gene Alias FLJ37870; JMY; junction mediating and regulatory protein, p53 cofactor; junction-mediating and regulatory protein; junction-mediating and -regulatory protein; LOC683593; p300 transcriptional cofactor JMY; similar to junction-mediating and regulatory protein; WAS protein homology region 2 domain containing 1-like 3; WHAMM2; WHDC1L3
Gene Symbols JMY
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human JMY. Recombinant protein control fragment (Product #RP-96900).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133746
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.