Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JNJ 0966
GREENER_CHOICE

Catalog No. 643910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-391-0 10 mg
64-395-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-391-0 Supplier Tocris Bioscience™ Supplier No. 6439/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pro-MMP9 activation inhibitor (IC50 = 440 nM)

JNJ 0966 is a pro-MMP9 activation inhibitor (IC50 = 440 nM). Inhibits the conversion of pro-MMP9 to active MMP9, without directly affecting MMP9 enzymatic activity. Exhibits selectivity for pro-MMP9 over pro-MMP1, 2 and 3. Inhibits cell invasion in in vitro assays. Reduces motor impairment in a mouse EAE model. Brain penetrant.

Specifications

Chemical Name or Material N-[2-[(2-Methoxyphenyl)amino]-4'-methyl[4,5'-bithiazol]-2'-yl]acetamide
CAS 315705-75-0
Quantity 10 mg
Molecular Formula C16H16N4O2S2
Formula Weight 360.45
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.