Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ JNJ 10191584 maleate
GREENER_CHOICESDP

Catalog No. 244150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-415-0 50 mg
24-411-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-415-0 Supplier R&D Systems™ Supplier No. 2441/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective H4 receptor antagonist; orally active

Highly selective histamine H4 receptor silent antagonist; binds with high affinity to the human H4 receptor (Ki = 26 nM). > 540-fold selective over the H3 receptor (Ki = 14.1 μM). Inhibits mast cell and eosinophil chemotaxis in vitro with IC50 values of 138 and 530 nM respectively. Orally active in vivo.

Chemical Identifiers

CAS 869497-75-6
Molecular Formula C17H21ClN4O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 394.81
Purity >99%
Molecular Weight (g/mol) 396.83
Chemical Name or Material JNJ 10191584 maleate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.