Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JNJ 17203212
GREENER_CHOICESDP

Catalog No. 336110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-611-0 10 mg
33-615-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-611-0 Supplier Tocris Bioscience™ Supplier No. 3361/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reversible, competitive and potent TRPV1 antagonist

Reversible, competitive and potent TRPV1 antagonist (pKi values are 6.5, 7.1 and 7.3 at rat, guinea pig and human TRPV1 respectively). Inhibits capsaicin- and H+-induced channel activation (pIC50 values are 6.32 and 7.23 respectively) and exhibits antitussive and analgesic activity in vivo.

Chemical Identifiers

CAS 821768-06-3
Molecular Formula C17H15F6N5O

Specifications

Quantity 10 mg
Formula Weight 419.32
Purity >99%
Molecular Weight (g/mol) 419.331
Chemical Name or Material JNJ 17203212
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.