Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JNJ 47965567
GREENER_CHOICESDP

Catalog No. 529910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-991-0 10 mg
52-995-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-991-0 Supplier Tocris Bioscience™ Supplier No. 5299/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective P2X7 antagonist; brain penetrant

JNJ 47965567 is a potent and selective P2X7 antagonist (pIC50 values are 8.3, 7.5 and 7.2 for human, mouse and rat receptors respectively). Selective over a panel of 50 other receptors, ion channels and transporters. Reduces BzATP -induced IL-1β release from monocytes in vitro and from rat brain in vivo. Attenuates amphetamine -induced hyperactivity in rats. Brain penetrant.

Specifications

Purity >98%
Quantity 10 mg
Product Type JNJ 47965567
Molecular Weight (g/mol) 488.64
Inhibitors JNJ 47965567

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.