Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JNJ 5207852 dihydrochloride
GREENER_CHOICE

Catalog No. 402010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-201-0 10 mg
40-205-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-201-0 Supplier Tocris Bioscience™ Supplier No. 4020/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity H3 receptor antagonist

High affinity histamine H3 receptor neutral antagonist (pKi values are 8.9 and 9.2 in rat and human respectively). Brain penetrant and orally active. Has 3- and 100-fold higher affinity than thioperamide (Cat. No. 0644) for rat and human H3 receptors respectively. Suppresses slow-wave sleep; exhibits wake-promoting effects in rodent arousal models.

Chemical Identifiers

CAS 398473-34-2
Molecular Formula C20H32N2O

Specifications

Quantity 10 mg
Formula Weight 389.4
Purity >99%
Molecular Weight (g/mol) 316.489
Chemical Name or Material JNJ 5207852 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.