Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JNJ 7777120
GREENER_CHOICE

Catalog No. 402125
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-212-5 25 mg
40-215 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-212-5 Supplier Tocris Bioscience™ Supplier No. 4021/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective H4 antagonist

JNJ 7777120 is a histamine H4 receptor antagonist; displays high affinity (Ki = 4.5 nM) and is >1000-fold selective for H4 over other histamine receptors.

Specifications

Chemical Name or Material 1-[(5-Chloro-1H-indol-2-yl)carbonyl]-4-methylpiperazine
CAS 459168-41-3
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C14H16ClN3O
Formula Weight Observed MW: 277.75
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.