Learn More
JNK Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | JNK |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5918
![]() ![]() |
Proteintech Group Inc
511511AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The MAPK8 protein can be phosphorylated and this antibody recognizes the 46 kDa and 55 kDa bands in western blot.
C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predomit forms are JNK1 and JNK2.Specifications
JNK | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P45983, P45984, P53779, Q61831, Q91Y86, Q9WTU6 | |
26414, 26419, 26420, 5599, 5601, 5602 | |
Peptide | |
Primary | |
-20°C | |
MAPK10 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
c Jun N terminal kinase 1, JNK 46, JNK1, JNK1A2, JNK21B1/2, MAP kinase 8, MAPK 8, MAPK8, PRKM8, SAPK1 | |
Mapk10, Mapk8, Mapk9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.