Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JNK2 Monoclonal Antibody (H.715.7)
GREENER_CHOICE

Catalog No. PIMA515183
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515183 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515183 Supplier Invitrogen™ Supplier No. MA515183
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone H.715.7
Concentration 278 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 50% glycerol, 0.15M NaCl, 100μg/mL BSA and <0.02% sodium azide; pH 7.5
Gene MAPK9
Gene Accession No. P45984, P49186, Q9WTU6
Gene Alias AI851083; c-Jun kinase 2; c-Jun N-terminal kinase 2; I79_012561; JNK/SAPK alpha; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; MAP kinase 9; MAPK 9; Mapk9; mitogen activated protein kinase 9; mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54a; p54-alpha; p54aSAPK; PRKM9; protein kinase, mitogen-activated 9; SAPK; SAPK1; SAPK1a; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Gene Symbols MAPK9
Host Species Rabbit
Immunogen Human JNK2/MBP fusion protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100764964, 26420, 50658, 5601
Target Species Hamster, Human, Mink, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.