Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JNK1 Monoclonal Antibody (M267)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547670
Description
This antibody detects a 46 kDa* protein corresponding to the apparent molecular mass of JNK1 on SDS-PAGE immunoblots of human A431 and HeLa, as well as rat PC12 cells.
MAPK8 (JNK1) is activated by the presence of misfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity. It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
Specifications
JNK1 | |
Monoclonal | |
~0.25 mg/mL | |
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
P45983, P49185, Q91Y86 | |
MAPK8 | |
Clone M267 was generated from a recombinant protein corresponding to amino acid residues in the C-terminal region of human JNK1. This sequence has high homology to rat and mouse JNK1, and has homology to similar regions in JNK2 and JNK3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
M267 | |
Unconjugated | |
MAPK8 | |
AI849689; c-Jun N-terminal kinase 1; FLJ12099; FLJ33785; JNK; JNK1; JNK1 beta1 protein kinase; jnk1a-1; JNK1A2; JNK21B1/2; JNK3; JNK3A; JNK-46; JUN N-terminal kinase; MAP kinase 10; MAP kinase 8; MAPK 10; MAPK 8; MAPK10; Mapk8; mapk8 protein; mapk8b; MGC50974; mitogen activated protein kinase 8; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 8b; MK10; OTTHUMP00000019552; p493F12; p54 gamma; p54bSAPK; Phospho-JNK; PRKM10; PRKM8; protein kinase JNK1; protein kinase mitogen-activated 8; SAPK gamma; SAPK1; SAPK1C; stress-activated protein kinase 1; stress-activated protein kinase 1c; Stress-activated protein kinase JNK1; zgc:112379 | |
Mouse | |
Protein A | |
RUO | |
116554, 26419, 5599 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction