Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JNK2 Antibody (004), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289620DL594
Description
JNK2 Monoclonal specifically detects JNK2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
JNK2 | |
Monoclonal | |
DyLight 594 | |
c-Jun kinase 2, c-Jun N-terminal kinase 2, EC 2.7.11, EC 2.7.11.24, JNK2A, JNK2BETA, JNK2JNK2ALPHA, JNK-55, Jun kinase, MAP kinase 9, MAPK 9, mitogen-activated protein kinase 9, p54a, p54aSAPK, PRKM9JNK2B, SAPK, Stress-activated protein kinase JNK2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human JNK2 (NP_002743.3; Met 1-Arg 424). | |
0.1 mL | |
Cancer, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
5601 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
4 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction