Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JNK2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298811AF594
Description
JNK2 Polyclonal specifically detects JNK2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
JNK2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
c-Jun kinase 2, c-Jun N-terminal kinase 2, EC 2.7.11, EC 2.7.11.24, JNK2A, JNK2BETA, JNK2JNK2ALPHA, JNK-55, Jun kinase, MAP kinase 9, MAPK 9, mitogen-activated protein kinase 9, p54a, p54aSAPK, PRKM9JNK2B, SAPK, Stress-activated protein kinase JNK2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human JNK2. | |
0.1 mL | |
Cancer, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
5601 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction