Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JNK2 Rabbit anti-Human, Clone: 11, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | JNK2 |
---|---|
Clone | 11 |
Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1-2 ul/mg of lysate, Immunohistochemistry-Paraffin 1:500-1:2500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
JNK2 Monoclonal specifically detects JNK2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
JNK2 | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1-2 ul/mg of lysate, Immunohistochemistry-Paraffin 1:500-1:2500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
c-Jun kinase 2, c-Jun N-terminal kinase 2, EC 2.7.11, EC 2.7.11.24, JNK2A, JNK2BETA, JNK2JNK2ALPHA, JNK-55, Jun kinase, MAP kinase 9, MAPK 9, mitogen-activated protein kinase 9, p54a, p54aSAPK, PRKM9JNK2B, SAPK, Stress-activated protein kinase JNK2 | |
Recombinant Human JNK2 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
11 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
0.2 um filtered solution in PBS | |
5601 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title