Learn More
Promega JNK3 Kinase Enzyme System, 1mg
Catalog No. PRV3822 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3822 1 mg
1 options
Catalog No. PRV3822 Supplier Promega Supplier No. V3822
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human JNK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. JNK3 is a member of the c-Jun N-terminal kinases (JNKs), which are part of the mitogen-activated protein (MAP) kinase family.
  • Easily Screen and Profile JNK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C