Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JNK3 Monoclonal Antibody (10E4A4)
GREENER_CHOICE

Catalog No. PIMA515403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515403 Supplier Invitrogen™ Supplier No. MA515403
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15403 targets MAPK10 in indirect ELISA, IF and WB applications and shows reactivity with Human and mouse samples. The MA5-15403 immunogen is purified recombinant fragment of human MAPK10 (aa28-233) expressed in E. Coli. MA5-15403 detects MAPK10 which has a predicted molecular weight of approximately 53kDa.

MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This protein is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). Through its phosphorylation and nuclear localization, this kinase plays regulatory roles in the signaling pathways during neuronal apoptosis. Beta-arrestin 2, a receptor-regulated MAP kinase scaffold protein, is found to interact with, and stimulate the phosphorylation of this kinase by MAP kinase kinase 4 (NhpK4). Cyclin-dependent kianse 5 can phosphorylate, and inhibit the activity of this kinase, which may be important in preventing neuronal apoptosis. Four alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10E4A4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene MAPK10
Gene Accession No. P53779, Q61831
Gene Alias C230008H04Rik; c-Jun N-terminal kinase 3; hypothetical protein LOC569698; jnk3; JNK3 alpha protein kinase; JNK3 beta1 protein kinase; JNK3 beta2 protein kinase; JNK3A; JNK3B1; JNK3B2; MAP kinase 10; MAP kinase p49 3F12; MAPK 10; MAPK10; mitogen activated protein kinase 10; mitogen-activated protein kinase 10; OTTHUMP00000161181; OTTHUMP00000161183; OTTHUMP00000218718; p493F12; p54-beta; p54bSAPK; PRKM10; SAPb; SAPK(beta); SAPK/Erk/kinase 2; SAPK1B; SAPK-beta; SAPKC; Serk2; Stress activated protein kinase beta; stress-activated protein kinase 1b; Stress-activated protein kinase JNK3; zgc:123234
Gene Symbols MAPK10
Host Species Mouse
Immunogen Purified recombinant fragment of human MAPK10 (aa28-233) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26414, 5602
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.