Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Jones and Foster Salt Mixture

Salt mixture for research animal diet.

Specifications

Product Type Powder
For Use With (Application) Research animal diet
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP290211002
View Documents
MP Biomedicals™
0290211002
2 kg N/A N/A N/A
MP290211010
View Documents
MP Biomedicals™
0290211010
10 kg N/A N/A N/A
MP290211050
View Documents
MP Biomedicals™
0290211050
50 kg N/A N/A N/A
Description

Description

  • Salt Mixture
  • For research animal diet
  • Powder
Specifications

Specifications

Powder
Research animal diet
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.