Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Josamycin, 98%, PROMO, Thermo Scientific Chemicals

Catalog No. J62245MFPM
Click to view available options
Quantity:
50 mg

Chemical Identifiers

CAS 16846-24-5
Molecular Formula C42H69NO15
Molecular Weight (g/mol) 828.01 g/mol
MDL Number MFCD00210320
InChI Key XJSFLOJWULLJQS-NGVXBBESSA-N
Synonym Leucomycin A3
PubChem CID 131849444
IUPAC Name (2S,3S,4R,6S)-6-{[(2R,3S,4R,5R,6S)-6-{[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-(acetyloxy)-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-6-yl]oxy}-4-(dimethylamino)-5-hydroxy-2-methyloxan-3-yl]oxy}-4-hydroxy-2,4-dimethyloxan-3-yl 3-methylbutanoate
SMILES CO[C@H]1[C@@H](CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]1O[C@H](C)[C@@H](O[C@H]2C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O2)[C@@H]([C@H]1O)N(C)C)OC(C)=O

Specifications

Type Josamycin
Physical Form Powder
Color White
Quantity 50 mg
Merck Index 14,5267
Solubility Information Soluble in ethanol
Percent Purity ≥98%
Chemical Name or Material Josamycin
EINECSNumber 240-871-6
RTECSNumber OH4725810
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.