Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JOSD1 Monoclonal Antibody (OTI3B11)
GREENER_CHOICE

Catalog No. PIMA525365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525365 Supplier Thermo Scientific Supplier No. MA525365
Add to Cart
Add to Cart

JOSD1 Monoclonal Antibody (OTI3B11)

JOSD1 Monoclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen JOSD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene JOSD1
Gene Accession No. Q15040
Gene Alias 1300006C06Rik; dJ508I15.2; Josd1; Josephin domain containing 1; josephin domain-containing 1; josephin domain-containing protein 1; Josephin-1; Josephin-1; josephin-1; JSPH1; KIAA0063; KIAA0063 gene product; mKIAA0063; RGD1305388; zgc:153682
Gene Symbols JOSD1
Host Species Mouse
Immunogen Full length human recombinant protein of JOSD1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9929
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.