Learn More
Invitrogen™ JOSD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551522
Description
Immunogen sequence: VFQDSNAFTR DTLQEIFQRL SPNTMVTPHK KSMLGNGNYD VNVIMAALQT KGYEAVWWDK RRDVGVIALT NVMGFIMNLP SSLCWGPLKL PLKRQHWICV REVGGAYYNL DSKLKMPEWI GGESELRKFL KHHLRGKNCE LLLVV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
Deubiquitinates monoubiquitinated probes (in vitro). When ubiquitinated, cleaves 'Lys-63'-linked and 'Lys-48'-linked poly-ubiquitin chains (in vitro), hence may act as a deubiquitinating enzyme. May increase macropinocytosis and suppress clathrin- and caveolae-mediated endocytosis. May enhance membrane dynamics and cell motility independently of its catalytic activity. [UniProt].
Specifications
| JOSD1 | |
| Polyclonal | |
| Unconjugated | |
| JOSD1 | |
| 1300006C06Rik; dJ508I15.2; Josd1; Josephin domain containing 1; josephin domain-containing 1; josephin domain-containing protein 1; Josephin-1; Josephin-1; josephin-1; JSPH1; KIAA0063; KIAA0063 gene product; mKIAA0063; RGD1305388; zgc:153682 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15040 | |
| JOSD1 | |
| Recombinant protein corresponding to Human JOSD1. Recombinant protein control fragment (Product #RP-91329). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.