Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JOSD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524098
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524098 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524098 Supplier Invitrogen™ Supplier No. PA524098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JOSD2 Polyclonal Antibody for Western Blot, Flow

The specific function of Josephin-2 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JOSD2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene JOSD2
Gene Accession No. Q8TAC2, Q9CR30
Gene Alias 1110007C05Rik; JOSD2; Josephin domain containing 2; josephin domain-containing protein 2; Josephin-2; SBBI54
Gene Symbols JOSD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 33-61 amino acids from the N-terminal region of human JOSD2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126119, 66124
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.