Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Jouberin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565106 Supplier Invitrogen™ Supplier No. PA565106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVAMYSDLPFK SPIRDISYHP FENMVAFCAF GQNEPILLYI YDFHVAQQEA EMFKRYNGTF PLPGIHQSQD ALCTCPKLPH QGSFQIDEFV HTESSSTKMQ LVKQ Highest antigen sequence identity to the following orthologs - mouse 88%, rat 87%.

This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Jouberin
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AHI1
Gene Accession No. Q8N157
Gene Alias 1700015F03Rik; Abelson helper integration site 1; abelson helper integration site 1 protein; abelson helper integration site 1 protein homolog; Ahi1; AHI-1; contatins SH3 and WD40 domains; D10Bwg0629e; dJ71N10.1; JBTS3; jouberin; ORF1; RP1-32B1.2
Gene Symbols AHI1
Host Species Rabbit
Immunogen Recombinant Human Jouberin. Recombinant protein control fragment (Product #RP-94850).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54806
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.