Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JPH2 Antibody (OTI1F5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272085H
Description
JPH2 Monoclonal specifically detects JPH2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
JPH2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
JPH2 | |
Full length recombinant protein of human JPH2 (NP_065166) produced in HEK293T cells. | |
0.1 mL | |
57158.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1F5 | |
Western Blot | |
JP2, junctophilin 2, junctophilin type 2, junctophilin-2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction