Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JPH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 405300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-530-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-530-0 Supplier Invitrogen™ Supplier No. 405300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JPH2 Polyclonal Antibody for IHC (F), IHC (P), Western Blot

This gene encodes a protein with a leucine-rich repeat and a calponin homology domain. Polymorphism in this gene may be associated with susceptibililty to knee osteoarthritis. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JPH2
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene JPH2
Gene Accession No. Q9ET78
Gene Alias 1110002E14Rik; CMH17; Jp2; JP-2; JP2NT; JPH2; jph2 {ECO:0000312; junctophilin 2; junctophilin type 2; Junctophilin-2; Junctophilin-2 N-terminal fragment; RGD:1305196}
Gene Symbols JPH2
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the mouse junctophilin-2 (JPH2, JP-2, junctophilin type 2) protein, which differs from rat by one non-conservative amino acid.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59091
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.