Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JPH3 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281726F
This item is not returnable.
View return policy
Description
JPH3 Polyclonal antibody specifically detects JPH3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
JPH3 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
FITC | |
CAGL237, junctophilin 3, junctophilin type 3 | |
JPH3 antibody was raised against a 18 amino acid synthetic peptide near the center of human JPH3. The immunogen is located within amino acids 410 - 460 of JPH3. Amino Acid Squence: PSFQHRENGLEYQRPKR | |
0.1 mL | |
Signal Transduction | |
57338 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction