Learn More
Invitrogen™ JPH3 Polyclonal Antibody

Description
A suggested positive control is Daudi cell lysate. PA5-20643 can be used with blocking peptide PEP-0763.
Specifications
Specifications
| Antigen | JPH3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | JPH3 |
| Gene Accession No. | Q8WXH2, Q9ET77 |
| Gene Alias | CAGL237; HDL2; JP3; JP-3; JPH3; junctophilin 3; junctophilin type 3; Junctophilin-3; TNRC22; trinucleotide repeat containing 22; Trinucleotide repeat-containing gene 22 protein |
| Gene Symbols | JPH3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.