Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JPH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111738 Supplier Invitrogen™ Supplier No. PA5111738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRRYSKGGAC RGLGDDHRPE DRGFGVQRLR SKAQNKENFR PASSAEPAVQ KLASLRLGGA EPRLLRWDLT F Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 67%.

Junctional complexes between the plasma membrane (PM) and endoplasmic/sarcoplasmic reticulum (ER/SR) are a common feature of all excitable cell types and mediate cross talk between cell surface and intracellular ion channels. Junctophilins (JPs) are important components of the junctional complexes. JPs are composed of a carboxy-terminal hydrophobic segment spanning the ER/SR membrane and a remaining cytoplasmic domain that shows specific affinity for the PM. Four JPs have been identified as tissue-specific subtypes derived from different genes: JPH1 is expressed in skeletal muscle, JPH2 is detected throughout all muscle cell types, and JPH3 and JPH4 are predominantly expressed in the brain. In the CNS, both JPH3 and JPH4 are expressed throughout neural sites and contribute to the subsurface cistern formation in neurons. Mice lacking both JPH3 and JPH4 subtypes exhibit serious symptoms such as impaired learning and memory and are accompanied by abnormal nervous functions. A repeat expansion in JPH3 is associated with Huntington disease-like 2. At least two isoforms of JPH3 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JPH3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JPH3
Gene Accession No. Q8WXH2
Gene Alias CAGL237; HDL2; JP3; JP-3; JPH3; junctophilin 3; junctophilin type 3; Junctophilin-3; TNRC22; trinucleotide repeat containing 22; Trinucleotide repeat-containing gene 22 protein
Gene Symbols JPH3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human JPH3. Recombinant protein control fragment (Product #RP-108081).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.