Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JPH4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144805
Description
Immunogen sequence: PSSPASSRQP WRPPACRSPL PPGGDQGPFS SPKAWPEEWG GAGAQAEELA GYEAEDEAGM QGPGPRDGSP LLGGCSDSSG SLREEE Antigen sequence identity: Mouse 92%; Rat 92%.
Junctional complexes between the plasma membrane (PM) and endoplasmic/sarcoplasmic reticulum (ER/SR) are a common feature of all excitable cell types and mediate cross talk between cell surface and intracellular ion channels. Junctophilins (JPs) are important components of the junctional complexes. JPs are composed of a carboxy-terminal hydrophobic segment spanning the ER/SR membrane and a remaining cytoplasmic domain that shows specific affinity for the PM. Four JPs have been identified as tissue-specific subtypes derived from different genes: JPH1 is expressed in skeletal muscle, JPH2 is detected throughout all muscle cell types, and JPH3 and JPH4 are predominantly expressed in the brain. In the CNS, both JPH3 and JPH4 are expressed throughout neural sites and contribute to the subsurface cistern formation in neurons. Mice lacking both JPH3 and JPH4 subtypes exhibit serious symptoms such as impaired learning and memory and are accompanied by abnormal nervous functions.
Specifications
JPH4 | |
Polyclonal | |
Unconjugated | |
JPH4 | |
9330157P13Rik; JP4; JP-4; Jph4; jph4 {ECO:0000312; JPHL1; junctophilin 4; junctophilin like 1; junctophilin-4; Junctophilin-like 1 protein; KIAA1831; RGD:1303170} | |
Rabbit | |
Affinity chromatography | |
RUO | |
84502 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JJ6 | |
JPH4 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 461-546. Recombinant protein control fragment (Product #RP-109941). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction