Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (+)-JQ1
GREENER_CHOICESDP

Catalog No. 449910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-991-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-991-0 Supplier Tocris Bioscience™ Supplier No. 4499/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective BET bromodomain inhibitor; cell permeable

(+)-JQ1 is a potent, high affinity, selective BET bromodomain inhibitor (IC50 values are 17.7, 32.6, 76.9 and 12942 nM for BRD2 (N-terminal (N)), BRD4 (C-terminal (C)), BRD4 (N) and CREBBP respectively; Kd values are 49, 59.5, 82, 90.1, 128 and 190 nM for BRD4 (N), BRD3 (N), BRD3 (C), BRD4 (C), BRD2 (N) and BRDT (N) respectively). (+)-JQ1 induces squamous differentiation in NUT midline carcinoma (NMC) cell lines and inhibits tumor growth in NMC xenograft models in vivo. (+)-JQ1 inhibits proliferation and induces autophagy in bladder cancer cells in vitro and in vivo. It also suppresses MYC gene expression and inhibits proliferation of lymphoma and leukemia cell lines. In human pulmonary microvasular endothelial cells (HPMEC), NF-κB activation, IL-6 and IL-8 expression and proliferation are inhibited by (+)-JQ1. The compound also inhibits transcription of ACE2 and TMPRSS2 genes in mouse lung tissue and prevents infection by SARS-CoV-2. In germ cells from male mice, (+)-JQ1 exhibits reversible contraceptive effects. (+)-JQ1 inhibits the BRD4-JUN-CCL2 axis in pancreatic cancer cells and improves survival by reducing macrophage recruitment.

Inactive Analog also available.

Carboxylic acid-functionalized (Cat. No. 6588) and click-activated (alkyne) (Cat. No. 6589) versions for PROTAC development also available.

Chemical Identifiers

CAS 1268524-70-4
Molecular Formula C23H25ClN4O2S

Specifications

Quantity 10 mg
Formula Weight 456.99
Purity >98%
Molecular Weight (g/mol) 456.989
Chemical Name or Material (+)-JQ1
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.