Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JRK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598370 Supplier Thermo Scientific Supplier No. PA598370
Add to Cart
Add to Cart

JRK Polyclonal Antibody

JRK Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen JRK
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene JRK
Gene Accession No. O75564, Q60976
Gene Alias jerky; jerky homolog; jerky protein; Jerky protein homolog; JH8; JRK; Jrk helix-turn-helix protein; Jrk homolog
Gene Symbols JRK
Host Species Rabbit
Immunogen Peptide sequence from Human Jerky protein homolog protein (465-483AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16469, 8629
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.