Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JRK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144048
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144048 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144048 Supplier Invitrogen™ Supplier No. PA5144048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human K562 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The JRK gene encodes a protein known as JRK (Jerky) homolog, which is implicated in various cellular processes, including DNA damage response and transcription regulation. JRK is conserved across multiple species and plays a role in the stability of neuronal functions and development. It is notably associated with neuropsychiatric disorders, where aberrations in its expression or function can contribute to conditions such as epilepsy and intellectual disabilities. The protein interacts with other cellular components to influence gene expression patterns, which are critical for maintaining normal neuronal activity. JRK is also studied for its potential involvement in cancer biology, where it may impact cell cycle regulation and genomic stability. Understanding the functions and mechanisms of JRK can provide insights into its roles in both normal physiological processes and disease states, particularly in the context of the nervous system and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JRK
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene JRK
Gene Accession No. O75564
Gene Alias jerky; jerky homolog; jerky protein; Jerky protein homolog; JH8; JRK; Jrk helix-turn-helix protein; Jrk homolog
Gene Symbols JRK
Host Species Rabbit
Immunogen E.coli-derived human JRK recombinant protein (Position: M1-Q257).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8629
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.