Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JRKL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561174
Description
Immunogen sequence: LQHTKGLENV TTENLEKWLE VDSTEPGYEV LTDSEIIRRA QGQADESSEN EEEEIELIPE KHINHAAALQ Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
The function of this gene has not yet been defined, however, the encoded protein shares similarity with the human (41% identical) and mouse (34% identical) jerky gene products. This protein may act as a nuclear regulatory protein. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, May 2012].
Specifications
| JRKL | |
| Polyclonal | |
| Unconjugated | |
| JRKL | |
| HHMJG; Human homolog of mouse jerky gene protein; jerky protein homolog-like; JRKL; JRK-like | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y4A0 | |
| JRKL | |
| Recombinant protein corresponding to Human JRKL. Recombinant protein control fragment (Product #RP-97526). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction