Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JSRP1 Monoclonal Antibody (6A9)
GREENER_CHOICE

Catalog No. PIMA524513
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524513 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524513 Supplier Invitrogen™ Supplier No. MA524513
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MSMTTRAWEE LDGGLGSCQA LEDHSALAET QEDRASATPR LADSGSVPHD SQVAEGPSVD TRPKKMEKEP AARGTPGTGK ERLKAGASPR SVPARKKAQT.

The junction between the transverse tubules (T-tubules) and the sarcoplasmic reticulum (SR) of skeletal muscle is called the triad. At the triad, dihydropyridine receptors (DHPR) of the T-tubule serve as voltage sensors in excitation-contraction coupling, while ryanodine receptors (RyR), the calcium release channels, exist in the membrane of the terminal cisternae of the SR. It is thought that during slow phase depolarization of the T-tubule, a third protein, triadin (95 kDa) transmits electrochemical signals to the SR through direct interaction with both DHPR and RyR. Another skeletal muscle specific 90 kDa protein has also been localized to the this junction. This protein is phosphorylated by an intrinsic protein kinase in triads suggesting that it may play an important regulatory or structural role in the skeletal muscle triad junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JSRP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6A9
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene JSRP1
Gene Accession No. Q96MG2
Gene Alias 2300003C06Rik; 2310032K21Rik; homolog of mouse skeletal muscle sarcoplasmic reticulum protein JP-45; JP45; JP-45; Jsrp1; Junctional sarcoplasmic reticulum protein 1; Junctional SR; junctional-face membrane protein of 45 kDa homolog; skeletal muscle sarcoplasmic reticulum protein JP-45
Gene Symbols JSRP1
Host Species Mouse
Immunogen JSRP1 (NP_653217.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126306
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.